7-Fluoro-2,3,4,9-tetrahydro-1H-β-carboline 95%

Warning

H27780-MC
AAH27780-MCEA 167.72 USD
AAH27780-MC
7-Fluoro-2,3,4,9-tetrahydro-1H-β-carboline 95%
7-Fluoro-2,3,4,9-tetrahydro-1H-beta-carboline
Formula: C₁₁H₁₁FN₂
MW: 190.22 g/mol
Melting Pt: 216…224 °C
Storage Temperature: Ambient
MDL Number: MFCD09264527
CAS Number: 177858-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR