Ethyl-2-chloro-6-methylisonicotinate 97%

Warning

Synonyms: Ethyl 2-chloro-6-methylisonicotinateEthyl 2-chloro-6-methylpyridine-4-carboxylate

H27778-03
AAH27778-03EA 94.88 USD
AAH27778-03
Ethyl-2-chloro-6-methylisonicotinate 97%
Ethyl 2-chloro-6-methylisonicotinate
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Boiling Pt: 107 °C/1.5 mm
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD09842151
CAS Number: 3998-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR