2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2-Dimethyltetrahydropyran-4-oneTetrahydro-2,2-dimethyl-4H-pyran-4-one

H27774-03
AAH27774-03EA 280.99 USD
AAH27774-03
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 60…64 °C/8 mm
Storage Temperature: Ambient
MDL Number: MFCD01549337
CAS Number: 1194-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR