5-Chloro-3-pyridineboronic acid 95%

Warning

Synonyms: (5-Chloropyridin-3-yl)boronic acid

H27756-03 H27756-MD H27756-06
AAH27756-03EA 457.36 USD
AAH27756-03 AAH27756-06 AAH27756-MD
5-Chloro-3-pyridineboronic acid 95%
5-Chloro-3-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798243
CAS Number: 872041-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR