3-Iodo-5-methylpyridin-2-ol 95%
|
Warning
|
Synonyms:
2-Hydroxy-3-iodo-5-methylpyridine
H27751-MD
H27751-03
AAH27751-MDEA
94.46
USD
AAH27751-MD
AAH27751-03
3-Iodo-5-methylpyridin-2-ol 95%
3-Iodo-5-methylpyridin-2-ol
Formula:
C₆H₆INO MW: 235.03 g/mol Melting Pt: 190…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037469 CAS Number: 313678-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...