(4-(Bromomethyl)phenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Bromomethyl)phenylacetic acid

H27744-06 H27744-03
AAH27744-03EA 42.29 USD
AAH27744-03 AAH27744-06
(4-(Bromomethyl)phenyl)acetic acid 97%
(4-(Bromomethyl)phenyl)acetic acid
Formula: (C₆H₄(CH₂Br))CH₂COOH
MW: 229.07 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00010632
CAS Number: 13737-36-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR