(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate 98%

Synonyms: (S)-1-tert-Butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate1-tert-butyl 2-methyl (2S)-4,4-difluoropyrrolidine-1,2-dicarboxylate

H27727-MD H27727-03
AAH27727-MDEA 82.88 USD
AAH27727-MD AAH27727-03
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate 98%
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate
Formula: C₁₁H₁₇F₂NO₄
MW: 265.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094918
CAS Number: 203866-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR