6,6'-Dibromo-3,3'-bipyridine tech. 90%, Technical Grade

Warning

Synonyms: 6,6'-Dibromo-3,3'-bipyridine

H27726-MC
AAH27726-MCEA 96.45 USD
AAH27726-MC
6,6'-Dibromo-3,3'-bipyridine tech. 90%, Technical Grade
6,6'-Dibromo-3,3'-bipyridine
Formula: C₁₀H₆Br₂N₂
MW: 313.98 g/mol
Melting Pt: ca 249 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD09037441
CAS Number: 147496-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR