N-Acetyl-DL-serine ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ac-DL-Ser-OH

H27717-06
AAH27717-06EA 401.99 USD
AAH27717-06
N-Acetyl-DL-serine ≥98%
N-Acetyl-DL-serine
Formula: C₅H₉NO₄
MW: 147.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037248
CAS Number: 97-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR