1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester 95%

Synonyms: 5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole

H27703-03 H27703-MD
AAH27703-MDEA 174.27 USD
AAH27703-MD AAH27703-03
1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester 95%
1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester
Formula: C₉H₁₅BN₂O₂
MW: 194.04 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD07777079
CAS Number: 1044851-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR