(R)-(+)-XylBINAP 98%
|
Warning
|
Synonyms:
(R)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl
, (R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl
H27680-ME
H27680-MC
AAH27680-MCEA
90.47
USD
AAH27680-MC
AAH27680-ME
(R)-(+)-XylBINAP 98%
(R)-(+)-XylBINAP
Formula:
C₅₂H₄₈P₂ MW: 734.90 g/mol Melting Pt: 201…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD01630821 CAS Number: 137219-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...