1-Methyl-4-cyano-4-(3-methoxyphenyl)piperidine 97%

Warning

H27677-03 H27677-06
AAH27677-06EA 183.15 USD
AAH27677-06 AAH27677-03
1-Methyl-4-cyano-4-(3-methoxyphenyl)piperidine 97%
1-Methyl-4-cyano-4-(3-methoxyphenyl)piperidine
Formula: C₁₄H₁₈N₂O
MW: 230.31 g/mol
Melting Pt: 43…47 °C
Storage Temperature: Ambient
MDL Number: MFCD05857265
CAS Number: 5460-79-7
EINECS: 226-742-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR