(R)-1-(2-Methoxyphenyl)ethylamine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-1-(2-Methoxyphenyl)ethanamine
H27669-03
H27669-06
AAH27669-03EA
161.99
USD
AAH27669-03
AAH27669-06
(R)-1-(2-Methoxyphenyl)ethylamine ≥99%, ee 98%, ChiPros®
(R)-1-(2-Methoxyphenyl)ethylamine
Formula:
C₉H₁₃NO MW: 151.21 g/mol Boiling Pt: 118 °C/17 mm Storage Temperature: Ambient |
MDL Number:
MFCD00671657 CAS Number: 68285-23-4 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...