2,3-Dichloro-4-pyridineboronic acid 95%

Warning

Synonyms: 2,3-Dichloropyridine-4-boronic acid2,3-Dichloro-4-pyridylboronic acid

H27668-MD H27668-03
AAH27668-03EA 231.99 USD
AAH27668-03 AAH27668-MD
2,3-Dichloro-4-pyridineboronic acid 95%
2,3-Dichloro-4-pyridineboronic acid
Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD04038756
CAS Number: 951677-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR