Methylamine 2 M in THF

Supplier: Thermo Scientific Chemicals
Danger

H27664-AE H27664-AU
AAH27664-AEEA 159 USD
AAH27664-AE AAH27664-AU
Methylamine 2 M in THF
Methylamine
Formula: CH₅N
MW: 31.06 g/mol
Density: 0.861
Flash Pt: <-34 °C(-29 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008104
CAS Number: 74-89-5
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR