2-Methyl-L-phenylalanine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Phe(2-Me)-OH

H27663-MC H27663-ME
AAH27663-MEEA 453.99 USD
AAH27663-ME AAH27663-MC
2-Methyl-L-phenylalanine 97%
2-Methyl-L-phenylalanine
Formula: C₁₀H₁₃NO₂
MW: 179.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044971
CAS Number: 80126-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR