4-Methyl-1-pentenylboronic acid 97%
|
Warning
|
H27655-03
H27655-MD
AAH27655-03EA
290.99
USD
AAH27655-03
AAH27655-MD
4-Methyl-1-pentenylboronic acid 97%
4-Methyl-1-pentenylboronic acid
Formula:
C₆H₁₃BO₂ Melting Pt: 99…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074680 CAS Number: 214907-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...