2-Benzyloxy-5-bromopyrimidine 95%
|
Warning
|
Synonyms:
2-(Benzyloxy)-5-bromopyrimidine
H27648-MD
H27648-03
AAH27648-MDEA
82.71
USD
AAH27648-MD
AAH27648-03
2-Benzyloxy-5-bromopyrimidine 95%
2-Benzyloxy-5-bromopyrimidine
Formula:
C₁₁H₉BrN₂O MW: 265.11 g/mol Melting Pt: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD07375140 CAS Number: 742058-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...