(S)-(-)-3-Chloro-α-methylbenzylamine ≥98%, ee 98%, ChiPros®

Danger

Synonyms: (S)-1-(3-Chlorophenyl)ethylamine(S)-1-(3-Chlorophenyl)ethanamine

H27637-03
AAH27637-03EA 99.3 USD
AAH27637-03
(S)-(-)-3-Chloro-α-methylbenzylamine ≥98%, ee 98%, ChiPros®
(S)-(-)-3-Chloro-α-methylbenzylamine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 112 °C/13 mm
Storage Temperature: Ambient
MDL Number: MFCD06761823
CAS Number: 68297-62-1
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR