(R)-(+)-Indoline-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27626-03
AAH27626-03EA 104.99 USD
AAH27626-03
(R)-(+)-Indoline-2-carboxylic acid 97%
(R)-(+)-Indoline-2-carboxylic acid
Formula: C₉H₉NO₂
Melting Pt: ca 177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00792496
CAS Number: 98167-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR