2-Ethoxycyclohexanone 98%

Warning

H27613-14 H27613-06 H27613-03
AAH27613-14EA 197.76 USD
AAH27613-14 AAH27613-03 AAH27613-06
2-Ethoxycyclohexanone 98%
2-Ethoxycyclohexanone
Formula: C₈H₁₄O₂
Boiling Pt: 85 °C/15 mm
Density: 1.002
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015377
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR