5,6'-Dibromo-3,3'-bipyridyl 90+%
|
Warning
|
Synonyms:
5,6'-Dibromo-3,3'-bipyridine
H27611-ME
H27611-MC
AAH27611-MEEA
362.97
USD
AAH27611-ME
AAH27611-MC
5,6'-Dibromo-3,3'-bipyridyl 90+%
5,6'-Dibromo-3,3'-bipyridyl
Formula:
C₁₀H₆Br₂N₂ Melting Pt: 201…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037423 CAS Number: 942206-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...