(R)-(-)-1-Boc-3-hydroxypyrrolidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-N-(tert-Butoxycarbonyl)-3-hydroxypyrrolidinetert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate

H27590-03 H27590-06
AAH27590-03EA 85.59 USD
AAH27590-03 AAH27590-06
(R)-(-)-1-Boc-3-hydroxypyrrolidine 98%
(R)-(-)-1-Boc-3-hydroxypyrrolidine
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01317838
CAS Number: 109431-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR