4-Benzyloxy-5-bromo-2-(1-pyrrolidinyl)pyrimidine 95%

Warning

H27587-03
AAH27587-03EA 158.73 USD
AAH27587-03
4-Benzyloxy-5-bromo-2-(1-pyrrolidinyl)pyrimidine 95%
4-Benzyloxy-5-bromo-2-(1-pyrrolidinyl)pyrimidine
Formula: C₁₅H₁₆BrN₃O
MW: 334.22 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD06798238
CAS Number: 885952-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR