(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

H27585-MD H27585-06 H27585-03
AAH27585-06EA 391 USD
AAH27585-06 AAH27585-MD AAH27585-03
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Formula: C₄₈H₄₀P₂
Melting Pt: 267…271 °C
Storage Temperature: Ambient
MDL Number: MFCD01311709
CAS Number: 153305-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR