1-Boc-4-cyanopiperidine 96%

Warning

Synonyms: N-BOC-Piperidine-4-carbonitriletert-Butyl 4-cyanopiperidine-1-carboxylate

H27580-03 H27580-MD H27580-06
AAH27580-MDEA 73.37 USD
AAH27580-MD AAH27580-03 AAH27580-06
1-Boc-4-cyanopiperidine 96%
1-Boc-4-cyanopiperidine
Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
Melting Pt: 45…48 °C
Storage Temperature: Ambient
MDL Number: MFCD01861223
CAS Number: 91419-52-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR