1,1'-Dibutylzirconocene dichloride ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(butylcyclopentadienyl)zirconium(IV) Dichloride
H27576-03
H27576-06
AAH27576-03EA
110.99
USD
AAH27576-03
AAH27576-06
1,1'-Dibutylzirconocene dichloride ≥98%
1,1'-Dibutylzirconocene dichloride
Formula:
C₁₈H₂₆Cl₂Zr MW: 404.53 g/mol Melting Pt: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672105 CAS Number: 73364-10-0 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...