Methyl-5-methyl-2-furoate 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Methyl 5-methylfuran-2-carboxylate
H27570-03
AAH27570-03EA
62.69
USD
AAH27570-03
Methyl-5-methyl-2-furoate 97%
Methyl 5-methyl-2-furoate
Formula:
C₇H₈O₃ MW: 140.14 g/mol Boiling Pt: 255 °C Density: 1.114 Flash Pt: 87 °C(189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04116374 CAS Number: 2527-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...