Isopropyldiphenylphosphine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27565-06
H27565-03
AAH27565-03EA
61.79
USD
AAH27565-03
AAH27565-06
Isopropyldiphenylphosphine 99%
Isopropyldiphenylphosphine
Formula:
C₁₅H₁₇P MW: 228.27 g/mol Boiling Pt: 165 °C/11 mm Melting Pt: 38…41 °C Density: 0.861 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015028 CAS Number: 6372-40-3 EINECS: 228-902-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...