3-Bromo-2,6-dimethoxypyridine 98%

Warning

H27564-06
AAH27564-06EA 94.12 USD
AAH27564-06
3-Bromo-2,6-dimethoxypyridine 98%
3-Bromo-2,6-dimethoxypyridine
Formula: C₇H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD08669938
CAS Number: 13445-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR