(S)-(-)-1-Boc-3-methylpiperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-tert-Butyl 3-methylpiperazine-1-carboxylate
, (3S)-3-Methylpiperazine, N1-BOC protected
H27534-06
H27534-03
AAH27534-06EA
258.99
USD
AAH27534-06
AAH27534-03
(S)-(-)-1-Boc-3-methylpiperazine 98%
(S)-(-)-1-Boc-3-methylpiperazine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Melting Pt: 38…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683204 CAS Number: 147081-29-6 UN: 3259 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...