Oxynitrox® S100, M.W. ∼ 2,250 ≥99% free radical

Warning

H27530-22 H27530-06 H27530-14
AAH27530-06EA 77.98 USD
AAH27530-06 AAH27530-22 AAH27530-14
Oxynitrox® S100, M.W. ∼ 2,250 ≥99% free radical
Oxynitrox® S100

Steers the selective oxidization of primary alcohols into aldehydes. Additionally, secondary alcohols can be converted into ketones

Storage Temperature: Ambient
CAS Number: 91993-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR