4-Methyl-D-phenylalanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Phe(4-Me)-OHH-p-Me-D-Phe-OH

H27518-03
AAH27518-03EA 217.99 USD
AAH27518-03
4-Methyl-D-phenylalanine 98%
4-Methyl-D-phenylalanine
Formula: C₁₀H₁₃NO₂
MW: 179.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152137
CAS Number: 49759-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR