3-Chlorocoumarin 97%
|
Warning
|
H27494-06
H27494-03
AAH27494-03EA
56.48
USD
AAH27494-03
AAH27494-06
3-Chlorocoumarin 97%
3-Chlorocoumarin
Formula:
C₉H₅ClO₂ Melting Pt: 121…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024073 CAS Number: 92-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...