5,2'-Dichloro-2,3'-bipyridyl 95%
|
Warning
|
Synonyms:
5,2'-Dichloro-2,3'-bipyridine
H27492-ME
H27492-MC
AAH27492-MEEA
321.9
USD
AAH27492-ME
AAH27492-MC
5,2'-Dichloro-2,3'-bipyridyl 95%
5,2'-Dichloro-2,3'-bipyridyl
Formula:
C₁₀H₆Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD09037433 CAS Number: 942206-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...