7-Hydroxy-3,4-dihydroquinolin-2(1H)-one 97%

Warning

Synonyms: 7-Hydroxyquinolinone

H27485-03
AAH27485-03EA 40.01 USD
AAH27485-03
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one 97%
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one
Formula: C₉H₉NO₂
MW: 163.17 g/mol
Melting Pt: 230…234 °C
Storage Temperature: Ambient
MDL Number: MFCD06410891
CAS Number: 22246-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR