5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine 95%

Warning

Synonyms: 5-Bromo-3-methyl-2-(methyl-pivaloylamino)pyridine

H27468-03 H27468-MD
AAH27468-03EA 267.51 USD
AAH27468-03 AAH27468-MD
5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine 95%
5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine
Formula: C₁₂H₁₇BrN₂O
MW: 285.18
Storage Temperature: Ambient
MDL Number: MFCD09037448
CAS Number: 245765-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR