2,2,4'-Tribromoacetophenone 97%
|
Danger
|
H27466-06
H27466-14
AAH27466-06EA
37.37
USD
AAH27466-06
AAH27466-14
2,2,4'-Tribromoacetophenone 97%
2,2,4'-Tribromoacetophenone
Formula:
C₈H₅Br₃O Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017858 CAS Number: 13195-79-4 EINECS: 236-161-0 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...