Polyethylene glycol methyl ether methacrylate, M.W. 5,000
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Polyethylene glycol monomethyl ether Methacrylate
, mPEG Methacrylate
, Methoxy PEG Methacrylate
046838-03
046838-MD
046838-06
AA46838-MDEA
34.99
USD
AA46838-MD
AA46838-06
AA46838-03
Polyethylene glycol methyl ether methacrylate, M.W. 5,000
Polyethylene glycol methyl ether methacrylate
Granular solid
Formula:
CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00241432 CAS Number: 26915-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...