Silicon ≥99.999% (metals basis), powder -325 mesh
Supplier: Alfa Aesar
|
Warning
|
46654-09
46654-18
AA46654-09EA
81.69
USD
AA46654-09
AA46654-18
Silicon ≥99.999% (metals basis), powder -325 mesh
Silicon
Formula:
Si Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...