Polyethylene glycol dimethanesulfonate, M.W. 8,000

46559-06 46559-MD 46559-03
AA46559-03EA 65.37 USD
AA46559-03 AA46559-MD AA46559-06
Polyethylene glycol dimethanesulfonate, M.W. 8,000
Polyethylene glycol dimethanesulfonate

Granular solid

Formula: CH₃SO₂(OCH₂CH₂)nOSO₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD03100947
CAS Number: 52410-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR