Polyethylene glycol dimethacrylate, M.W. 1,000
|
Warning
|
Synonyms:
PEGDMA
, PEG dimethacrylate
46534-03
46534-MD
46534-06
AA46534-06EA
436.9
USD
AA46534-06
AA46534-03
AA46534-MD
Polyethylene glycol dimethacrylate, M.W. 1,000
Polyethylene glycol dimethacrylate
Granular solid
Formula:
(C₂H₄O)nC₈H₁₀O₃ Storage Temperature: Freezer |
MDL Number:
MFCD00014932 CAS Number: 25852-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...