Polyethylene glycol methyl ether methacrylate, M.W. 13,000

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Polyethylene glycol monomethyl ether MethacrylatemPEG MethacrylateMethoxy PEG Methacrylate

046513-03 046513-06 46513-MD
AA46513-06EA 397.99 USD
AA46513-06 AA46513-MD AA46513-03
Polyethylene glycol methyl ether methacrylate, M.W. 13,000
Polyethylene glycol methyl ether methacrylate

Granular solid

Formula: CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃
Storage Temperature: Refrigerator
MDL Number: MFCD00241432
CAS Number: 26915-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR