Polyester acrylate/methacrylate PEAM-1769

Supplier: Thermo Scientific Chemicals
Warning

046351-22 046351-14
AA46351-22EA 294.99 USD
AA46351-22 AA46351-14
Polyester acrylate/methacrylate PEAM-1769
Polyester acrylate/methacrylate

Amber liquid


Applications: Recommended for use as a base resin or an additive to reduce stress

MW: 645 g/mol
Storage Temperature: Refrigerator
CAS Number: 921213-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR