Poly(styrene sulfonic acid) sodium salt, M.W. 75,000

Supplier: Alfa Aesar

Synonyms: Poly(sodium-p-styrenesulfonate)Poly(sodium-p-styrenesulphonate)

45851-30 45851-18 45851-0B
AA45851-30EA 104 USD
AA45851-30 AA45851-18 AA45851-0B
Poly(styrene sulfonic acid) sodium salt, M.W. 75,000
Poly(styrene sulfonic acid) sodium salt

Powder

Formula: [-CH₂CH(C₆H₄)SO₃Na)-]n
Storage Temperature: Ambient
MDL Number: MFCD00084449
CAS Number: 25704-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR