Zinc oxide, HiFUEL® A310 for sulfur removal, based sulfur removal material

Supplier: Alfa Aesar
Warning

Synonyms: Zinci oxidumZinc (II) oxide

45469-A1 45469-36 45469-A4
AA45469-A1EA 153.21 USD
AA45469-A1 AA45469-36 AA45469-A4
Zinc oxide, HiFUEL® A310 for sulfur removal, based sulfur removal material
Zinc oxide

Spheres, 2-5mm dia

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR