PTFE, sheets, Thickness 1.6 mm (0.063 in)

Supplier: Alfa Aesar

Synonyms: TeflonPolytetrafluoroethylene

45194-HB
AA45194-HBEA 73.2 USD
AA45194-HB
PTFE, sheets, Thickness 1.6 mm (0.063 in)
PTFE
Formula: (CF₂CF₂)n
Storage Temperature: Ambient
MDL Number: MFCD00084455
CAS Number: 9002-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR