Polyethylene, rods, low density, Diameter 12.7 mm (0.5 in)
45179-BV
AA45179-BVEA
27.65
USD
AA45179-BV
Polyethylene, rods, low density, Diameter 12.7 mm (0.5 in)
Polyethylene
Formula:
(CH₂CH₂)n Storage Temperature: Ambient |
MDL Number:
MFCD00084424 CAS Number: 64754-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...