Polyethylene, sheets, high density, Thickness 1.6 mm (0.063 in)

45155-HB
AA45155-HBEA 25.6 USD
AA45155-HB
Polyethylene, sheets, high density, Thickness 1.6 mm (0.063 in)
Polyethylene (CAS 9002-88-4)
Formula: (CH₂CH₂)n
Storage Temperature: Ambient
MDL Number: MFCD00084423
CAS Number: 9002-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR