Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Copper, alloy 101, foil, Oxygen-Free High Conductivity (OFHC), Thickness 0.81 mm (0.032 in)

Supplier: Alfa Aesar

45147-HB 45147-GT
AA45147-HBEA 142.23 USD InStock
AA45147-HB AA45147-GT
Copper, alloy 101, foil, Oxygen-Free High Conductivity (OFHC), Thickness 0.81 mm (0.032 in)
Copper, alloy 101


MW: 63.55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
EINECS: 231-159-6
Merck Index: 13,02545
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty